Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 1, 2 | 286 | ||
NM_033624.2 | NP_296373.1 | 1, 2 | 286 | |||
XM_017019037.1 | XP_016874526.1 | 1, 2 | 291 | |||
XM_017019038.1 | XP_016874527.1 | 1, 2 | 291 | |||
GenBank mRNA | AB020682.1 | - | 1, 2 | 248 | ||
AF174601.1 | - | 1, 2 | 247 | |||
AK001699.1 | - | 1, 2 | 270 | |||
AK308141.1 | - | 2, 3 | 193 | |||
AK314573.1 | - | 1, 2 | 260 | |||
BC034045.1 | - | 2, 3 | 213 | 4815947 | ||
BC042333.1 | - | 2, 3 | 186 | 5195515 | ||
BC091496.1 | - | 1, 2 | 245 | |||
BI550121.1 | - | 1, 2 | 297 |