Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135911.1 | NP_001129383.1 | 5 | 713 | ||
NM_052880.4 | NP_443112.2 | 6 | 792 | |||
GenBank mRNA | AF528080.1 | - | 6 | 737 | ||
AK074688.1 | - | 6 | 738 | |||
AK093768.1 | - | 3 | 1352 | |||
AK226089.1 | - | 6 | 672 | |||
AK299397.1 | - | 5 | 667 | |||
AK300990.1 | - | 7 | 650 | |||
BC011049.2 | - | 6 | 737 | 4180134 | ||
BC041903.1 | - | 4 | 1089 | |||
BE299406.1 | - | 4 | 420 | |||
CR456340.1 | - | 6 | 632 | |||
GU135609.1 | - | 4 | 416 |