Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000495.4 | NP_000486.1 | 45 | 4386 | ||
NM_033380.2 | NP_203699.1 | 47 | 4404 | |||
XM_011530849.2 | XP_011529151.2 | 48 | 4152 | |||
XM_017029259.1 | XP_016884748.1 | 47 | 4143 | |||
XM_017029260.1 | XP_016884749.1 | 46 | 4134 | |||
XM_017029263.1 | XP_016884752.1 | 32 | 3230 | |||
GenBank mRNA | AK307900.1 | - | 5 | 616 | ||
BC151846.1 | - | 47 | 4366 | 8860442 | ||
KM282618.1 | - | 48 | 4377 | |||
M31115.1 | - | 14 | 1365 | |||
M58526.1 | - | 42 | 3862 |