Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033518.3 | NP_277053.2 | 11 | 930 | ||
XM_005272694.3 | XP_005272751.2 | 10 | 920 | |||
XM_005272695.4 | XP_005272752.3 | 10 | 922 | |||
XM_005272697.2 | XP_005272754.2 | 10 | 873 | |||
XM_005272698.4 | XP_005272755.2 | 10 | 811 | |||
XM_006724569.3 | XP_006724632.1 | 11 | 930 | |||
XM_017029960.1 | XP_016885449.1 | 10 | 842 | |||
XM_017029961.1 | XP_016885450.1 | 10 | 814 | |||
GenBank mRNA | AF276889.1 | - | 10 | 1595 | ||
AF318358.1 | - | 7 | 1041 | |||
AK094788.1 | - | 10 | 849 | |||
BC019246.2 | - | 10 | 780 | 3506044 | ||
BC027721.2 | - | 6 | 471 |