Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033540.2 | NP_284941.2 | 9 | 1059 | ||
XM_005247596.3 | XP_005247653.2 | 9 | 1017 | |||
XM_011512963.2 | XP_011511265.1 | 6 | 645 | |||
GenBank mRNA | AB043588.1 | - | 8 | 1024 | ||
AB043589.1 | - | 9 | 1024 | |||
AF329637.1 | - | 8 | 933 | |||
AK000700.1 | - | 9 | 1016 | |||
AK314306.1 | - | 9 | 1016 | |||
BC020661.1 | - | 9 | 1025 | |||
BC040557.1 | - | 9 | 1059 | 5270347 | ||
CR627379.1 | - | 6 | 709 | |||
U95822.1 | - | 5 | 522 |