Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033549.4 | NP_291027.3 | 4, 5 | 1894 | ||
NM_201627.2 | NP_963921.1 | 4, 5 | 1894 | |||
XM_006714929.2 | XP_006714992.1 | 4, 5 | 1892 | |||
XM_006714930.2 | XP_006714993.1 | 4, 5 | 1892 | |||
XM_006714931.3 | XP_006714994.1 | 4, 5 | 1892 | |||
XM_006714932.3 | XP_006714995.1 | 4, 5 | 1892 | |||
GenBank mRNA | AB063180.1 | - | 4, 5 | 920 | ||
AB100366.1 | - | 4, 5 | 1157 | |||
AB100367.1 | - | 4, 5 | 1157 | |||
AF258579.1 | - | 1 | 50 | |||
AK027601.1 | - | 4, 5 | 1867 | |||
AK074052.1 | - | 2 | 3335 | |||
AK307695.1 | - | 2 | 822 | |||
AL137593.1 | - | 1 | 1027 | |||
AL832960.1 | - | 4 | 771 | |||
AX775789.1 | - | 4, 5 | 633 | |||
AX775791.1 | - | 4, 5 | 633 | |||
AX775793.1 | - | 4, 5 | 1867 | |||
AX775797.1 | - | 4, 5 | 925 | |||
AX775799.1 | - | 4, 5 | 1867 | |||
AX775801.1 | - | 4 | 1867 | |||
AX775803.1 | - | 4 | 925 | |||
BC018765.1 | - | 4, 5 | 925 | |||
BC090953.1 | - | 4, 5 | 1437 | 6017628 |