Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002211.3 | NP_002202.2 | 10, 11 | 1498 | ||
NM_033668.2 | NP_391988.1 | 9, 10 | 1277 | |||
NM_133376.2 | NP_596867.1 | 10, 11 | 1413 | |||
GenBank mRNA | AK291697.1 | - | 10, 11 | 1350 | ||
BC020057.1 | - | 10, 11 | 1342 | 3924411 | ||
BM791591.1 | - | 1, 2 | 114 | |||
BX537407.1 | - | 10, 11 | 1352 |