Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127215.1 | NP_001120687.1 | 7 | 1873 | ||
NM_001127216.1 | NP_001120688.1 | 7 | 1866 | |||
NM_005263.3 | NP_005254.2 | 7 | 1959 | |||
XM_005270749.3 | XP_005270806.1 | 7 | 2315 | |||
XM_011541245.2 | XP_011539547.1 | 7 | 2801 | |||
XM_011541246.2 | XP_011539548.1 | 7 | 2077 | |||
GenBank mRNA | AK123009.1 | - | 7 | 2028 | ||
BC032751.1 | - | 7 | 1843 | 5585794 | ||
U67369.1 | - | 7 | 1976 |