Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 1385 | ||
NM_001322924.1 | NP_001309853.1 | 5 | 1313 | |||
NM_052862.3 | NP_443094.3 | 7 | 1431 | |||
GenBank mRNA | AF545852.1 | - | 7 | 1575 | ||
AK025425.1 | - | 2 | 770 | |||
AK124221.1 | - | 1 | 1153 | |||
AL833211.1 | - | 5 | 1121 | |||
BC013186.1 | - | 2 | 480 | |||
BC072399.1 | - | 2 | 723 | |||
BC098426.1 | - | 2 | 596 | |||
BC101536.1 | - | 7 | 1268 | 8069042 | ||
BC101562.1 | - | 7 | 1268 | 8069068 | ||
BC143428.1 | - | 6 | 1178 | 9051936 |