Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052885.3 | NP_443117.3 | 10 | 1958 | ||
XM_011537847.2 | XP_011536149.1 | 9 | 2058 | |||
XM_017018764.1 | XP_016874253.1 | 10 | 1431 | |||
XM_017018765.1 | XP_016874254.1 | 10 | 2069 | |||
XM_017018766.1 | XP_016874255.1 | 9 | 1216 | |||
GenBank mRNA | AJ315644.1 | - | 10 | 1958 | ||
BC117117.1 | - | 10 | 1902 | 40125668 | ||
BC117119.1 | - | 10 | 1902 | 40125670 |