Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286250.1 | NP_001273179.1 | 10 | 2882 | ||
NM_001286251.1 | NP_001273180.1 | 9 | 2783 | |||
NM_001286254.2 | NP_001273183.1 | 9 | 1948 | |||
NM_001323252.1 | NP_001310181.1 | 12 | 3037 | |||
NM_001323253.1 | NP_001310182.1 | 9 | 2702 | |||
NM_001323254.1 | NP_001310183.1 | 8 | 2486 | |||
NM_001323257.1 | NP_001310186.1 | 10 | 2774 | |||
NM_001323260.1 | NP_001310189.1 | 11 | 2986 | |||
NM_001323261.1 | NP_001310190.1 | 9 | 2500 | |||
NM_001323262.1 | NP_001310191.1 | 7 | 2212 | |||
NM_001323263.1 | NP_001310192.1 | 9 | 2504 | |||
NM_001323264.1 | NP_001310193.1 | 8 | 2288 | |||
NM_052904.3 | NP_443136.2 | 11 | 2990 | |||
NR_104421.1 | 9 | 2047 | ||||
NR_136579.1 | 8 | 1777 | ||||
NR_136580.1 | 10 | 2263 | ||||
XM_005266813.4 | XP_005266870.1 | 9 | 2395 | |||
XM_017010228.1 | XP_016865717.1 | 8 | 2179 | |||
XM_017010229.1 | XP_016865718.1 | 9 | 2352 | |||
GenBank mRNA | AB067487.1 | - | 9 | 2404 | ||
AF351610.1 | - | 9 | 1878 | |||
AK055292.1 | - | 11 | 2876 | |||
BC031830.1 | - | 8 | 1628 | |||
BC047491.1 | - | 9 | 1828 |