Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052957.4 | NP_443189.1 | 4, 5 | 644 | ||
XM_006724716.3 | XP_006724779.1 | 4, 5 | 197 | |||
XM_011531063.2 | XP_011529365.1 | 3, 4 | 464 | |||
XM_011531064.1 | XP_011529366.1 | 4, 5 | 247 | |||
XM_017029965.1 | XP_016885454.1 | 5, 6 | 298 | |||
GenBank mRNA | AJ311392.1 | - | 3, 4 | 145 | ||
AK097433.1 | - | 4, 5 | 258 | |||
AK127607.1 | - | 3, 4 | 680 | |||
BC136259.1 | - | 4, 5 | 212 | 9020246 |