Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300764.1 | NP_001287693.1 | 3, 4 | 466 | ||
NM_001300766.1 | NP_001287695.1 | 3, 4 | 466 | |||
NM_052965.3 | NP_443197.1 | 3, 4 | 466 | |||
XM_006711148.1 | XP_006711211.1 | 3, 4 | 415 | |||
GenBank mRNA | AF288394.1 | - | 3, 4 | 441 | ||
AK098196.1 | - | 3, 4 | 364 | |||
BC022030.1 | - | 3, 4 | 411 | 4824025 | ||
BI821111.1 | - | 3, 4 | 391 | |||
BP326115.1 | - | 3, 4 | 391 |