Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006166.3 | NP_006157.1 | 6, 7 | 713 | ||
XM_005268908.4 | XP_005268965.1 | 7, 8 | 777 | |||
XM_005268909.4 | XP_005268966.1 | 7, 8 | 760 | |||
XM_005268910.4 | XP_005268967.1 | 7, 8 | 1432 | |||
GenBank mRNA | AK291934.1 | - | 6, 7 | 645 | ||
BC005316.1 | - | 6, 7 | 662 | 3680937 | ||
BC005317.1 | - | 6, 7 | 668 | 3997299 | ||
BC007035.1 | - | 6, 7 | 640 | 3996244 | ||
L06145.1 | - | 4, 5 | 512 | |||
X59710.1 | - | 6, 7 | 617 |