Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037293.2 | NP_001032370.1 | 7 | 527 | ||
NM_053016.5 | NP_443749.5 | 7 | 741 | |||
GenBank mRNA | AJ312216.1 | - | 7 | 599 | ||
AK095003.1 | - | 7 | 493 |
Gene Symbol: PALM2-AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ312217.1 | - | 7 | 595 |