Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053053.3 | NP_444281.1 | 2 | 315 | ||
GenBank mRNA | AK290964.1 | - | 2 | 132 | ||
AK291922.1 | - | 2 | 255 | |||
AK309250.1 | - | 2 | 183 | |||
BC015401.2 | - | 2 | 140 | 4182025 | ||
CN399642.1 | - | 2 | 317 |