Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRASLS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146728.1 | NP_001140200.1 | 1, 2 | 330 | ||
NM_054108.3 | NP_473449.1 | 1, 2 | 330 | |||
XM_011544753.1 | XP_011543055.1 | 1, 2 | 63 | |||
GenBank mRNA | AB298804.1 | - | 1, 2 | 156 | ||
AJ416558.1 | - | 1, 2 | 317 | |||
AK302312.1 | - | 1, 2 | 291 | |||
DB460110.1 | - | 1, 2 | 332 |