Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004813.2 | NP_004804.1 | 3 | 521 | ||
NM_057174.2 | NP_476515.1 | 3 | 521 | |||
XM_011520474.1 | XP_011518776.1 | 3 | 260 | |||
GenBank mRNA | AB016531.1 | - | 3 | 246 | ||
AF118240.1 | - | 3 | 453 | |||
BC000467.2 | - | 3 | 227 | 2822827 | ||
BC004356.1 | - | 3 | 219 | 2822311 | ||
CN285513.1 | - | 3 | 528 |