Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304347.1 | NP_001291276.1 | 4 | 537 | ||
NM_001304348.1 | NP_001291277.1 | 3 | 236 | |||
NM_058164.3 | NP_477512.1 | 4 | 563 | |||
GenBank mRNA | AL535795.3 | - | 4 | 565 | ||
AL538120.3 | - | 4 | 530 | |||
BC011361.1 | - | 4 | 408 | 4213407 | ||
BI907480.1 | - | 3 | 234 | |||
BX406452.2 | - | 4 | 551 | |||
BX437233.2 | - | 3 | 235 |