Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003818.2 | NP_001003818.1 | 8 | 1326 | ||
NM_001198644.1 | NP_001185573.1 | 7 | 836 | |||
NM_001198645.1 | NP_001185574.1 | 6 | 740 | |||
NM_058166.4 | NP_477514.1 | 8 | 1127 | |||
GenBank mRNA | AF220030.1 | - | 8 | 1115 | ||
AK023210.1 | - | 5 | 378 | |||
AK027664.1 | - | 8 | 1119 | |||
AK290172.1 | - | 8 | 1326 | |||
AK293295.1 | - | 6 | 569 | |||
AK298301.1 | - | 7 | 665 | |||
AK316178.1 | - | 8 | 793 | |||
AK316289.1 | - | 8 | 921 | |||
BC047564.1 | - | 8 | 1305 | 4825118 | ||
BC065575.1 | - | 8 | 852 | 4609084 | ||
CR749260.1 | - | 9 | 1022 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CX760618.1 | - | 5 | 490 |