Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278270.1 | NP_001265199.1 | 3 | 618 | ||
NM_006354.3 | NP_006345.1 | 3 | 1224 | |||
NM_133480.2 | NP_597814.1 | 3 | 1224 | |||
GenBank mRNA | AF069733.1 | - | 2 | 401 | ||
AK000228.1 | - | 3 | 628 | |||
AK292264.1 | - | 3 | 618 | |||
AL117487.1 | - | 3 | 887 | |||
BC009240.2 | - | 3 | 949 | 3009860 | ||
BC013433.1 | - | 3 | 887 | 3897746 | ||
CR533543.1 | - | 2 | 401 | |||
DB032623.1 | - | 4 | 486 |