Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052937.3 | NP_443169.2 | 2 | 601 | ||
XM_005251146.2 | XP_005251203.1 | 2 | 601 | |||
XM_017013017.1 | XP_016868506.1 | 1 | 242 | |||
GenBank mRNA | AK056952.1 | - | 2 | 451 | ||
BC032670.1 | - | 1 | 346 | 5539318 |