Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258411.2 | NP_001245340.1 | 9 | 983 | ||
NM_001258412.2 | NP_001245341.1 | 8 | 878 | |||
NM_001258413.2 | NP_001245342.1 | 9 | 929 | |||
NM_052944.4 | NP_443176.2 | 9 | 939 | |||
XM_005255080.2 | XP_005255137.1 | 10 | 967 | |||
XM_005255082.4 | XP_005255139.1 | 4 | 373 | |||
XM_006721015.2 | XP_006721078.1 | 8 | 947 | |||
XM_011545725.2 | XP_011544027.2 | 9 | 1110 | |||
XM_017022900.1 | XP_016878389.1 | 10 | 1283 | |||
XM_017022901.1 | XP_016878390.1 | 10 | 1993 | |||
XM_017022902.1 | XP_016878391.1 | 10 | 1122 | |||
XM_017022903.1 | XP_016878392.1 | 11 | 1132 | |||
XM_017022904.1 | XP_016878393.1 | 11 | 1302 | |||
XM_017022905.1 | XP_016878394.1 | 9 | 1178 | |||
XM_017022906.1 | XP_016878395.1 | 10 | 1143 | |||
GenBank mRNA | AF292385.1 | - | 8 | 707 | ||
AJ305237.1 | - | 8 | 725 | |||
AK125267.1 | - | 11 | 1055 | |||
AK295427.1 | - | 8 | 747 | |||
AK298373.1 | - | 8 | 724 | |||
AK303691.1 | - | 9 | 851 | |||
AY044906.1 | - | 9 | 939 | |||
BC057780.1 | - | 9 | 798 | 5287366 | ||
U41898.1 | - | 6 | 491 |