Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317808.1 | NP_001304737.1 | 2 | 501 | ||
NM_001317809.1 | NP_001304738.1 | 2 | 614 | |||
NM_001317810.1 | NP_001304739.1 | 3 | 767 | |||
NM_001317811.1 | NP_001304740.1 | 3 | 621 | |||
NM_052953.3 | NP_443185.1 | 2 | 768 | |||
XM_005264849.3 | XP_005264906.1 | 2 | 1739 | |||
XM_005264850.4 | XP_005264907.1 | 2 | 1124 | |||
XM_011533332.2 | XP_011531634.1 | 3 | 614 | |||
XM_017005675.1 | XP_016861164.1 | 2 | 1228 | |||
GenBank mRNA | AF396933.2 | - | 2 | 765 | ||
AY358282.1 | - | 2 | 354 | |||
BC040656.1 | - | 3 | 769 | 4798356 | ||
BC067784.1 | - | 2 | 768 | 5265009 | ||
BC087849.1 | - | 3 | 621 | 30383500 | ||
BI917966.1 | - | 2 | 614 |