Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320768.1 | NP_001307697.1 | 4 | 933 | ||
NM_052954.3 | NP_443186.2 | 4 | 930 | |||
NR_135472.1 | 5 | 1100 | ||||
GenBank mRNA | AF401639.1 | - | 4 | 535 | ||
AK054581.1 | - | 4 | 810 | |||
AK223576.1 | - | 4 | 650 | |||
AK304124.1 | - | 5 | 980 | |||
AL833200.1 | - | 4 | 741 | |||
AY061853.1 | - | 4 | 475 | |||
BC036761.1 | - | 4 | 718 | 5588436 | ||
KC843341.1 | - | 3 | 370 | |||
KF154022.1 | - | 5 | 702 |