Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300764.1 | NP_001287693.1 | 4 | 472 | ||
NM_001300766.1 | NP_001287695.1 | 4 | 472 | |||
NM_052965.3 | NP_443197.1 | 4 | 472 | |||
NR_023349.2 | 5 | 589 | ||||
NR_125335.1 | 5 | 552 | ||||
XM_006711148.1 | XP_006711211.1 | 4 | 421 | |||
GenBank mRNA | AF288394.1 | - | 4 | 447 | ||
AK098196.1 | - | 4 | 370 | |||
AK307241.1 | - | 5 | 480 | |||
BC022030.1 | - | 4 | 417 | 4824025 | ||
BI821111.1 | - | 4 | 397 | |||
BP326115.1 | - | 4 | 397 | |||
BU195337.1 | - | 5 | 524 |