Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079536.1 | NP_001073004.1 | 3 | 542 | ||
NM_001297668.1 | NP_001284597.1 | 2 | 855 | |||
NM_053017.4 | NP_443750.2 | 2 | 855 | |||
XM_005252779.3 | XP_005252836.1 | 2 | 531 | |||
XM_006718136.3 | XP_006718199.1 | 3 | 426 | |||
XM_011519879.2 | XP_011518181.1 | 3 | 576 | |||
GenBank mRNA | AK313531.1 | - | 2 | 534 | ||
AY358466.1 | - | 2 | 480 | |||
BC014577.1 | - | 2 | 546 | 3934249 | ||
Y16835.1 | - | 3 | 545 |