Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006593.3 | NP_006584.1 | 5 | 1493 | ||
GenBank mRNA | AK226050.1 | - | 2 | 3543 | ||
AK297168.1 | - | 2 | 457 | |||
AK297438.1 | - | 4 | 661 | |||
AK299533.1 | - | 4 | 393 | |||
AK312567.1 | - | 5 | 1268 | |||
AK315865.1 | - | 4 | 388 | |||
AK316161.1 | - | 4 | 524 | |||
BC029289.1 | - | 5 | 1254 | |||
BC104844.1 | - | 5 | 1421 | 8143847 | ||
BC113418.1 | - | 5 | 1421 | 8322470 | ||
U49250.1 | - | 5 | 1200 |