Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304347.1 | NP_001291276.1 | 6 | 928 | ||
NM_001304348.1 | NP_001291277.1 | 5 | 627 | |||
NM_058164.3 | NP_477512.1 | 6 | 954 | |||
GenBank mRNA | AF131839.1 | - | 6 | 791 | ||
BC011361.1 | - | 6 | 799 | 4213407 | ||
BI907480.1 | - | 5 | 625 | |||
BQ069013.1 | - | 6 | 735 | |||
BX437233.2 | - | 5 | 626 |