Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005861.2 | NP_001005861.1 | 15 | 2608 | ||
NM_002958.3 | NP_002949.2 | 15 | 2599 | |||
XM_017007016.1 | XP_016862505.1 | 10 | 2105 | |||
GenBank mRNA | AB209405.1 | - | 5 | 4935 | ||
S59184.1 | - | 15 | 2728 |