Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006735.3 | NP_006726.1 | 2 | 1543 | ||
GenBank mRNA | BC130571.1 | - | 2 | 1297 | 40146602 | |
BC136500.1 | - | 2 | 1297 | 9020489 |