Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256409.1 | NP_001243338.1 | 9 | 1622 | ||
NM_052940.4 | NP_443172.1 | 8 | 1588 | |||
XM_006710327.3 | XP_006710390.1 | 9 | 1444 | |||
XM_006710328.3 | XP_006710391.1 | 9 | 1957 | |||
GenBank mRNA | AK075201.1 | - | 8 | 1550 | ||
AK090881.1 | - | 9 | 1871 | |||
AK095767.1 | - | 9 | 1541 | |||
BC013101.1 | - | 8 | 1427 | 3918785 |