Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053282.4 | NP_444512.2 | 3 | 370 | ||
GenBank mRNA | AF256653.1 | - | 3 | 248 | ||
AF403479.1 | - | 3 | 248 | |||
AF484964.1 | - | 3 | 346 | |||
AK097162.1 | - | 3 | 297 | |||
AK314743.1 | - | 3 | 317 | |||
BC022407.1 | - | 3 | 358 | 4717041 |