Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANTXR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145794.1 | NP_001139266.1 | 3, 4 | 1069 | ||
NM_001286780.1 | NP_001273709.1 | 3, 4 | 232 | |||
NM_001286781.1 | NP_001273710.1 | 3, 4 | 769 | |||
NM_058172.5 | NP_477520.2 | 3, 4 | 1069 | |||
XM_011531587.2 | XP_011529889.1 | 3, 4 | 232 | |||
XM_017007711.1 | XP_016863200.1 | 3, 4 | 769 | |||
GenBank mRNA | AB209913.1 | - | 3, 4 | 657 | ||
AK055636.1 | - | 2, 3 | 120 | |||
AY040326.1 | - | 3, 4 | 446 | |||
AY233452.1 | - | 3, 4 | 309 | |||
BC114374.1 | - | 3, 4 | 181 |