Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324086.1 | NP_001311015.1 | 3, 4 | 434 | ||
NM_001324410.1 | NP_001311339.1 | 3, 4 | 335 | |||
NM_001324412.1 | NP_001311341.1 | 2, 3 | 245 | |||
NM_001324413.1 | NP_001311342.1 | 3, 4 | 335 | |||
NM_058192.2 | NP_478072.1 | 3, 4 | 434 | |||
GenBank mRNA | AL514840.3 | - | 3, 4 | 285 | ||
BC011783.2 | - | 2, 3 | 299 | 3937184 | ||
BG686882.1 | - | 3, 4 | 433 | |||
BM763460.1 | - | 3, 4 | 421 | |||
BX358716.2 | - | 2, 3 | 230 | |||
CN311779.1 | - | 3, 4 | 293 |