Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080652.3 | NP_542383.1 | 3, 4 | 539 | ||
GenBank mRNA | AK290878.1 | - | 3, 4 | 517 | ||
AK313954.1 | - | 3, 4 | 475 | |||
AY358547.1 | - | 3, 4 | 523 | |||
BC008043.2 | - | 3, 4 | 496 | 3838280 | ||
BC019884.1 | - | 3, 4 | 486 | 4991416 | ||
BP274550.1 | - | 3, 4 | 540 |