Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080658.1 | NP_542389.1 | 4 | 500 | ||
XM_017018549.1 | XP_016874038.1 | 5 | 691 | |||
XM_017018550.1 | XP_016874039.1 | 5 | 725 | |||
XM_017018551.1 | XP_016874040.1 | 5 | 646 | |||
GenBank mRNA | AF359506.1 | - | 3 | 419 | ||
AY032593.1 | - | 480 | ||||
AY040761.1 | - | 4 | 480 | |||
AY169233.1 | - | 4 | 500 | |||
BC008689.1 | - | 4 | 500 | 3853707 |