Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242366.2 | NP_001229295.1 | 2 | 487 | ||
NM_080669.5 | NP_542400.2 | 2 | 487 | |||
XM_005277786.3 | XP_005277843.1 | 2 | 487 | |||
XM_017024110.1 | XP_016879599.1 | 3 | 446 | |||
GenBank mRNA | AK054669.1 | - | 2 | 485 | ||
AK295883.1 | - | 2 | 443 | |||
BC010691.1 | - | 2 | 476 | 3872267 | ||
DA188147.1 | - | 2 | 487 | |||
DB121588.1 | - | 2 | 303 |