Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080670.3 | NP_542401.1 | 3 | 1354 | ||
GenBank mRNA | AF445025.1 | - | 2 | 2607 | ||
AJ420598.1 | - | 1 | 367 | |||
AK289378.1 | - | 1 | 708 | |||
AL833972.1 | - | 2 | 1073 | |||
BC009906.2 | - | 3 | 1084 | 2961449 |