Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004638.3 | NP_004629.3 | 2 | 375 | ||
NM_080686.2 | NP_542417.2 | 2 | 338 | |||
XM_017011274.1 | XP_016866763.1 | 1 | 90 | |||
GenBank mRNA | AB210034.1 | - | 2 | 324 | ||
AK302756.1 | - | 2 | 299 | |||
AK309774.1 | - | 2 | 299 | |||
BC032134.2 | - | 2 | 259 | |||
BC042295.1 | - | 2 | 301 | 5555847 | ||
BC060668.1 | - | 2 | 262 | 6471026 | ||
DC345271.1 | - | 2 | 338 | |||
M33509.1 | - | 2 | 190 |