Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324412.1 | NP_001311341.1 | 3, 4 | 362 | ||
NM_001324413.1 | NP_001311342.1 | 4, 5 | 452 | |||
NM_001324414.1 | NP_001311343.1 | 4, 5 | 508 | |||
NM_001324415.1 | NP_001311344.1 | 4, 5 | 561 | |||
NM_058192.2 | NP_478072.1 | 4, 5 | 551 | |||
GenBank mRNA | BC011783.2 | - | 3, 4 | 416 | 3937184 | |
BC012951.2 | - | 51 | ||||
BC036279.2 | - | 51 | ||||
BG686882.1 | - | 4, 5 | 550 | |||
BX358716.2 | - | 3, 4 | 347 | |||
CN311777.1 | - | 4, 5 | 465 | |||
CN311779.1 | - | 4, 5 | 410 | |||
CX786721.1 | - | 4, 5 | 511 |