Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320974.1 | NP_001307903.1 | 3 | 943 | ||
NM_001320975.1 | NP_001307904.1 | 3 | 943 | |||
NM_001320976.1 | NP_001307905.1 | 3 | 943 | |||
NM_004376.6 | NP_004367.2 | 3 | 943 | |||
NM_078470.5 | NP_510870.1 | 3 | 943 | |||
XM_006717634.3 | XP_006717697.1 | 3 | 408 | |||
XM_011539298.2 | XP_011537600.1 | 3 | 408 | |||
XM_017015715.1 | XP_016871204.1 | 3 | 944 | |||
GenBank mRNA | AF026850.1 | - | 3 | 377 | ||
AF044323.1 | - | 3 | 377 | |||
AK291654.1 | - | 3 | 372 | |||
AK298864.1 | - | 3 | 372 | |||
BC002382.2 | - | 3 | 342 | 2961532 | ||
BC013403.2 | - | 3 | 342 | 2961532 | ||
BC078161.1 | - | 3 | 354 | 6150426 | ||
BT007129.1 | - | 3 | 326 | |||
BX537557.1 | - | 3 | 943 |