Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017926.2 | NP_001017926.1 | 3 | 2997 | ||
NM_007222.4 | NP_009153.3 | 3 | 2902 | |||
NR_037873.1 | 3 | 2833 | ||||
GenBank mRNA | AF106862.1 | - | 1 | 2316 | ||
AF195766.1 | - | 3 | 2665 | |||
AK025236.1 | - | 1 | 391 | |||
AK289532.1 | - | 3 | 2833 | |||
BC040481.1 | - | 3 | 2722 | 5271344 |