Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020857.2 | NP_065908.1 | 2, 3 | 566 | ||
XM_017022447.1 | XP_016877936.1 | 2, 3 | 586 | |||
GenBank mRNA | AB040908.1 | - | 2, 3 | 541 | ||
AF308802.1 | - | 2, 3 | 541 | |||
AL713725.1 | - | 2, 3 | 566 | |||
BC001513.2 | - | 2, 3 | 393 | 2822780 |