Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012296.3 | NP_036428.1 | 2 | 297 | ||
NM_080491.2 | NP_536739.1 | 2 | 353 | |||
XM_006718753.2 | XP_006718816.1 | 3 | 398 | |||
XM_011545408.2 | XP_011543710.1 | 2 | 248 | |||
GenBank mRNA | AB011143.1 | - | 2 | 297 | ||
AB018413.1 | - | 2 | 251 | |||
BC131711.1 | - | 2 | 255 | 40122341 | ||
BC152459.1 | - | 2 | 297 | 8862595 | ||
BY799914.2 | - | 2 | 302 | |||
DB636423.1 | - | 3 | 430 |