Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080625.3 | NP_542192.2 | 7 | 1103 | ||
XM_006723706.3 | XP_006723769.1 | 7 | 1166 | |||
XM_011528566.1 | XP_011526868.1 | 8 | 1230 | |||
XM_011528567.2 | XP_011526869.1 | 8 | 1230 | |||
XM_011528568.2 | XP_011526870.1 | 9 | 1356 | |||
XM_011528569.1 | XP_011526871.1 | 8 | 1230 | |||
GenBank mRNA | AK057090.1 | - | 8 | 883 | ||
AK125588.1 | - | 3 | 506 | |||
BC030254.1 | - | 7 | 1095 | 5182496 | ||
BC032455.1 | - | 3 | 293 |