Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080652.3 | NP_542383.1 | 4 | 545 | ||
XM_017007437.1 | XP_016862926.1 | 3 | 383 | |||
GenBank mRNA | AK172748.1 | - | 3 | 1725 | ||
AK290878.1 | - | 4 | 523 | |||
AK313954.1 | - | 4 | 481 | |||
AY358547.1 | - | 4 | 529 | |||
BC008043.2 | - | 4 | 502 | 3838280 | ||
BC019884.1 | - | 4 | 492 | 4991416 | ||
BP274550.1 | - | 4 | 546 | |||
BX647928.1 | - | 2 | 2417 |