Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012101.3 | NP_036233.2 | 9 | 2864 | ||
XM_005271490.1 | XP_005271547.1 | 9 | 2041 | |||
XM_011542729.1 | XP_011541031.1 | 10 | 2990 | |||
XM_011542732.1 | XP_011541034.1 | 10 | 2987 | |||
XM_011542733.1 | XP_011541035.1 | 10 | 2143 | |||
XM_017017453.1 | XP_016872942.1 | 10 | 2299 | |||
XM_017017454.1 | XP_016872943.1 | 9 | 2197 | |||
GenBank mRNA | AF230388.1 | - | 9 | 2864 | ||
AF230389.1 | - | 11 | 2042 | |||
AK093404.1 | - | 5 | 1934 | |||
AK122720.1 | - | 10 | 2680 | |||
AL832678.1 | - | 2 | 2806 | |||
BC010644.1 | - | 9 | 2810 | 3855016 | ||
BC017352.2 | - | 9 | 2803 | 5013158 | ||
BX648072.1 | - | 1 | 3585 | |||
L24203.1 | - | 9 | 2864 |