Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163771.1 | NP_001157243.1 | 2, 3 | 468 | ||
NM_080679.2 | NP_542410.2 | 2, 3 | 468 | |||
NM_080680.2 | NP_542411.2 | 2, 3 | 468 | |||
NM_080681.2 | NP_542412.2 | 2, 3 | 468 | |||
XM_017010250.1 | XP_016865739.1 | 3, 4 | 733 | |||
GenBank mRNA | BC053886.1 | - | 2, 3 | 454 |