Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004638.3 | NP_004629.3 | 6 | 764 | ||
NM_080686.2 | NP_542417.2 | 6 | 727 | |||
XM_017011274.1 | XP_016866763.1 | 5 | 479 | |||
GenBank mRNA | AB210034.1 | - | 6 | 713 | ||
AK302756.1 | - | 6 | 688 | |||
AK309774.1 | - | 6 | 688 | |||
BC032134.2 | - | 6 | 648 | |||
BC042295.1 | - | 6 | 690 | 5555847 | ||
BC060668.1 | - | 6 | 651 | 6471026 | ||
M33509.1 | - | 6 | 579 |